Diethyl(isopropyl)silanol ≥95.0% (by GC)
Catalog # TCIAD2746-5G
Supplier: TCI
CAS Number:
Specifications
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:146.31 g/mol
CAS Number:1158830-31-9
Formula:C₇H₁₈OSi
MDL Number:MFCD03093625