5,6-Dihydro-1,4-dithiin-2,3-dicarboximide ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:24519-85-5
:
Formula:C₆H₅NO₂S₂
MDL Number:MFCD01632185
:187.23 g/mol
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Neutralization titration):min. 97.0 %
- Melting point:216.0 to 219.0 deg-C
- Solubility in THF:very faint turbidity
Specification Test Results
Frequently Bought Together