Order Entry
Sweden
ContactUsLinkComponent
DSP (Dithiobis(succinimidyl propionate)) ≥97.0% (by total nitrogen basis) cross-linking reagent
Irritant
DSP (Dithiobis(succinimidyl propionate)) ≥97.0% (by total nitrogen basis) cross-linking reagent
Supplier:  TCI
Certificates
Irritant
DSP (Dithiobis(succinimidyl propionate)) ≥97.0% (by total nitrogen basis) cross-linking reagent
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (with Total Nitrogen):
    min. 97.0 %
  • Melting point:
    128.0 to 133.0 deg-C

Specification Test Results

Related Information