Order Entry
Sweden
ContactUsLinkComponent
5-(1,3-Dioxolan-2-yl)-2-furaldehyde ≥95.0% (by GC)
5-(1,3-Dioxolan-2-yl)-2-furaldehyde ≥95.0% (by GC)
Catalog # TCIAD2418-1G
Supplier:  TCI
CAS Number:  
5-(1,3-Dioxolan-2-yl)-2-furaldehyde ≥95.0% (by GC)
Catalog # TCIAD2418-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 95.0 %
  • Melting point:
    40.0 to 45.0 deg-C

Specification Test Results

About this item

 :
 :168.15 g/mol
Storage Temperature:Refrigerator
CAS Number:117953-13-6
Formula:C₈H₈O₄
MDL Number:MFCD00671521