Order Entry
Sweden
ContactUsLinkComponent
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
trans-N-(6,6-Dimethyl-2-hepten-4-ynyl)-N-methyl-1-naphthylmethylamine hydrochloride ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information