Order Entry
Sweden
ContactUsLinkComponent
 
 
 
 
1,3-Divinyl-1,1,3,3-tetramethyldisilazane ≥95.0% (by GC)
 :  TCI
 
undefined
 
 
 
1,3-Divinyl-1,1,3,3-tetramethyldisilazane ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

Frequently Bought Together