sec-Butyl ether (DL and meso mixture) ≥97.0% (by GC) stabilised
Supplier: TCI
Certificates
About this item
CAS Number:6863-58-7
:
Formula:C₈H₁₈O
MDL Number:MFCD00039921
:130.23 g/mol
Boiling Pt:121°C
Flash Pt:26°C
EINECS:229-961-6
UN:1149
ADR:3,III
:Stabilised with hydroquinone (HQ)
:GHS02
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 % (total of isomer)