3,5-Dimethyl-3-heptene (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Catalog # TCIAD1263-5ML
Supplier: TCI
CAS Number:
Specifications
- Pk:5 mL
- Seq:2210
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
Specification Test Results
About this item
:
:126.24 g/mol
Flash Pt:23°C
UN:3295
ADR:3,III
:GHS02
:GHS08
Formula:C₉H₁₈
MDL Number:MFCD00027049