
4,4'-Dimethyldiphenylamine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:620-93-9
:
Formula:C₁₄H₁₅N
MDL Number:MFCD00059317
:197.2756 g/mol
Boiling Pt:330°C
EINECS:210-659-8
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:78.0 to 81.0 deg-C
- Solubility in Methanol:almost transparency