Chloro-IB-MECA ≥98.0% (by HPLC)
: TCI
:163042-96-4
:
:C₁₈H₁₈ClIN₆O₄
:MFCD01861178
:544.73 g/mol
:Refrigerator
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:207.0 to 211.0 deg-C
- Elemental analysis (Nitrogen):14.70 to 16.20 %