Curcumol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4871-97-0
:
Formula:C₁₅H₂₄O₂
MDL Number:MFCD00272163
:236.36 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:141.0 to 145.0 deg-C
- Specific rotation [a]20/D:-36.0 to -39.5 deg (C=1, CHCL3)