Order Entry
Sweden
ContactUsLinkComponent
 
6-Chloro-7-hydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by GC)
 :  TCI
 
6-Chloro-7-hydroxy-4-methyl-2H-chromen-2-one ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %