
5-Chloro-2,3-diphenylpyrazine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:41270-66-0
:
Formula:C₁₆H₁₁ClN₂
MDL Number:MFCD00234892
:266.73 g/mol
Boiling Pt:145°C
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:125.0 to 129.0 deg-C