
6-(2-Chloroacetyl)-2H-1,4-benzoxazin-3(4H)-one ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:26518-76-3
:
Formula:C₁₀H₈ClNO₃
MDL Number:MFCD01321312
:225.63 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 97.0 %
- Melting point:228.0 to 232.0 deg-C