
2-Chloro-5-methylpyrimidine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:22536-61-4
:
Formula:C₅H₅ClN₂
MDL Number:MFCD09260903
:128.56 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:92.0 to 96.0 deg-C