Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(3-Chlorophenyl)acetone
(3-Chlorophenyl)acetone ≥95.0% (by GC)
(3-Chlorophenyl)acetone ≥95.0% (by GC)
:
TCI
(3-Chlorophenyl)acetone ≥95.0% (by GC)
:
TCI
:
14123-60-5
:
:
C₉H₉ClO
:
MFCD00082872
:
168.62 g/mol
:
99°C
Appearance:
White to Yellow to Green clear liquid
Purity (GC):
min. 95.0 %