trans-1,3-Cyclohexanediamine ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:26883-70-5
:
Formula:C₆H₁₄N₂
:114.19 g/mol
Boiling Pt:198°C
Density:0.97
UN:2735
ADR:8,III
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 95.0 %
- Purity (Nonaqueous Titration):min. 97.0 %