0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
2-Cyano-4-methylpyridine
2-Cyano-4-methylpyridine ≥98.0% (by GC)
2-Cyano-4-methylpyridine ≥98.0% (by GC)
:
TCI
2-Cyano-4-methylpyridine ≥98.0% (by GC)
:
TCI
:
1620-76-4
:
:
C₇H₆N₂
:
MFCD00128868
:
118.14 g/mol
:
148°C
:
3439
:
6.1,III
:
GHS06
Appearance:
White to Gray to Brown powder to crystal
Purity (GC):
min. 98.0 %
Melting point:
86.0 to 90.0 deg-C