
Chlorobis(ethylene)rhodium(I) dimer ≥95.0% (by titrimetric analysis)
Catalog # TCIAC2461-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (Argentmetric Titration):min. 95.0 %
- Rhodium:50.30 to 55.60 %
Specification Test Results
About this item
:
:388.93 g/mol
EINECS:235-145-0
:GHS07
Storage Temperature:Freezer
CAS Number:12081-16-2
Formula:C₈H₁₆Cl₂Rh₂
MDL Number:MFCD00013206