Order Entry
Sweden
ContactUsLinkComponent
 
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate ≥98.0% (by titrimetric analysis)
 :  TCI
 
(1R,2R)-(+)-1,2-Cyclohexanediamine-L-tartrate ≥98.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +10.0 to +13.0 deg (C=2.5, H2O)