
(R)-2-(((Benzyloxy)carbonyl)amino)succinic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:78663-07-7
:
Formula:C₁₂H₁₃NO₆
MDL Number:MFCD00063182
:267.2 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Solubility in hot Water:almost transparency
- Melting point:115.0 to 119.0 deg-C
- Specific rotation [a]20/D:-8.0 to -11.0 deg (C=7, AcOH)