1-Bromo-9,9'-spirobi[9H-fluorene] ≥98.0% (by GC)
Leverantör: TCI
Certifikat
Om denna produkt
CAS-nummer:1450933-18-2
:
Formel:C₂₅H₁₅Br
MDL-nummer:MFCD31556229
:395.30 g/mol
Specifikationstestresultat
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:212.0 to 216.0 deg-C