N,N'-Bis[3-(dimethylamino)propyl]perylene-3,4,9,10-tetracarboxylic diimide ≥95.0% (by total nitrogen basis)
: TCI
: DAPER, 2,9-Bis[3-(dimethylamino)propyl]anthra[2,1,9-def:6,5,10-d'e'f']diisoquinoline-1,3,8,10(2H,9H)-tetrone
:117901-97-0
:
:C₃₄H₃₂N₄O₄
:MFCD03791075
:560.65 g/mol
:300°C
- Environmentally Preferable:
- Appearance:Light yellow to Amber to Dark green powder to crystal
- Purity (with Total Nitrogen):min. 95.0 %