Order Entry
Sweden
Orders LinkContactUsLinkComponent
Bromotriethylsilane ≥96.0% (by GC)

Bromotriethylsilane ≥96.0% (by GC)

5 g
Catalog # TCIAB5890-5G
Flammable
Corrosive
Supplier:   TCI
Bromotriethylsilane ≥96.0% (by GC)
Catalog # TCIAB5890-5G
Supplier:  TCI
CAS Number:  1112-48-7
Bromotriethylsilane ≥96.0% (by GC)

5 g

CAS Number:  1112-48-7
Price
8 400,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Triethylbromosilane
  • CAS Number:1112-48-7
  • Formula:C₆H₁₅BrSi
  • MDL Number:MFCD00000049
  • Molecular Weight:195.17 g/mol
  • Boiling Pt:163°C
  • Flash Pt:53°C
  • EINECS:214-191-5
  • UN:2920
  • ADR:8,II
  •  :Density: 1.135 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Appearance:
    White to Almost white clear liquid
  • Purity (GC):
    min. 96.0 %

Specifications

  • Pk:
    5 g

Specifications