Order Entry
Sweden
ContactUsLinkComponent
 
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol ≥98.0% (by GC)
 :  TCI
 
2-[[tert-Butyl(dimethyl)silyl]oxy]ethanol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %