0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Bradford's reagent
Bradford's reagent for protein determination, ready-to-use
Bradford's reagent for protein determination, ready-to-use
TCIAB5702-500ML
:
TCI
:
Bradford's reagent for protein determination, ready-to-use
TCIAB5702-500ML
:
TCI
:
Förp.:
500 mL
Seq:
2725
Appearance:
Green to Dark green to Dark blue clear liquid
Protein determination test:
to pass test
:
:
GHS07
:
GHS08
:
Kylskåp