Order Entry
Sweden
ContactUsLinkComponent
1,8-Bis(maleimido)-3,6-dioxaoctane ≥98.0% (by HPLC)
1,8-Bis(maleimido)-3,6-dioxaoctane ≥98.0% (by HPLC)
Leverantör:  TCI
Certifikat
1,8-Bis(maleimido)-3,6-dioxaoctane ≥98.0% (by HPLC)
Leverantör:  TCI

Specifikationstestresultat

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Elemental analysis (Nitrogen):
    8.50 to 9.50 %
  • Melting point:
    96.0 to 100.0 deg-C

Specifikationstestresultat