tert-Butyl ((1R,2R)-2-aminocyclohexyl)carbamate ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:146504-07-6
:
Formula:C₁₁H₂₂N₂O₂
MDL Number:MFCD08726023
:214.31 g/mol
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:114.0 to 118.0 deg-C