Order Entry
Sweden
ContactUsLinkComponent
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98.0% (by HPLC)
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98.0% (by HPLC)
Supplier:  TCI
Certificates
undefined
(R,R)-(-)-N,N'-Bis(3,5-di-tert-butylsalicylidene)-1,2-cyclohexanediaminomanganese(III) chloride ≥98.0% (by HPLC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Orange to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Chelometric Titration):
    min. 96.0 %

Specification Test Results