Order Entry
Sweden
ContactUsLinkComponent
 
 
1-Butyl-3-methylimidazolium dicyanamide ≥96.0% (by HPLC, total nitrogen)
 :  TCI
 
undefined
 
1-Butyl-3-methylimidazolium dicyanamide ≥96.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    Colorless to Red to Green clear liquid
  • Purity (HPLC):
    min. 96.0 % (total of Ion-Pair)
  • Purity (with Total Nitrogen):
    min. 96.0 %