
1-Boc-indole-2-boronic acid (contains varying amounts of Anhydride)
Catalog # TCIAB5145-1G
Supplier: TCI
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (Neutralization titration):97.0 to 108.0 %
Specification Test Results
About this item
:
:261.08 g/mol
Melting Pt:105°C
:GHS07
Storage Temperature:Freezer
CAS Number:213318-44-6
Formula:C₁₃H₁₆BNO₄
MDL Number:MFCD02093045