Order Entry
Sweden
ContactUsLinkComponent
Methyl undecanoate
Methyl undecanoate
Catalog # ROTH6844.1
Supplier:  Roth Carl
CAS Number:  
Methyl undecanoate
Catalog # ROTH6844.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :200.32 g/mol
EINECS:217-053-2
CAS Number:1731-86-8
Formula:C₁₂H₂₄O₂
MDL Number:MFCD00008957