Bexarotene ≥96.0% (by HPLC)
Katalog # TCIAB4798-100MG
Supplier: TCI
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (HPLC):min. 96.0 area%
- Melting point:220.0 to 225.0 deg-C
Specification Test Results
About this item
:
:348.486 g/mol
Merck Index:14,01194
:GHS08
Storage Temperature:Freezer
CAS Number:153559-49-0
Formula:C₂₄H₂₈O₂
MDL Number:MFCD00932428