Order Entry
Sweden
ContactUsLinkComponent
 
1,3-Bis(4-methoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)
 :  TCI
 
1,3-Bis(4-methoxyphenyl)urea ≥98.0% (by HPLC, total nitrogen)
 :  TCI

 

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %
  • Melting point:
    239.0 to 243.0 deg-C

 

Frequently Bought Together