
Bis(isothiocyanato)bis(2,2'-bipyridyl-4,4'-dicarboxylato)ruthenium(II) ≥85.0% (by HPLC)
Catalog # TCIAB4372-200MG
Supplier: TCI
CAS Number:
Specifications
- Pk:200 mg
- Seq:2530
Specifications
Specification Test Results
- Appearance:Gray to Dark purple to Black powder to crystal
- Purity (HPLC):min. 85.0 area%
- Purity (Neutralization titration):min. 85.0 %
Specification Test Results
About this item
:
:705.64 g/mol
:GHS08
CAS Number:141460-19-7
Formula:C₂₆H₁₆N₆O₈RuS₂
MDL Number:MFCD11042213