0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
Methyl stearate
Methyl stearate
Methyl stearate
ACCUSFA011N
:
AccuStandard
:
Methyl stearate
ACCUSFA011N
:
AccuStandard
:
Pack type:
Glass bottle for solids
Pk:
100 mg
Seq:
2440
:
:
298.51 g/mol
:
12,08959
:
355 °C (1013 hPa)
:
37…39 °C
:
1.007 g/cm³ (20 °C)
:
Ambient
:
203-990-4
:
112-61-8
:
CH₃(CH₂)₁₆CO₂CH₃
:
MFCD00009005