(2R,5R)-(+)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:390766-89-9
:
Formula:C₁₅H₂₂N₂O
MDL Number:MFCD08276842
:246.35 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Optical purity (LC):min. 98.0 ee%
- Melting point:98.0 to 102.0 deg-C