
Bromodimethylsulphonium bromide ≥98.0% (by titrimetric analysis)
: TCI
:50450-21-0
:
:C₂H₆Br₂S
:MFCD10000904
:221.94 g/mol
:82°C
:Freezer
:3261
:8,III
:GHS05
- Appearance:Light yellow to Brown powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %