
3-Benzyloxy-1-propanol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:4799-68-2
:
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00029659
:166.22 g/mol
Boiling Pt:120°C
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %