Order Entry
Sweden
ContactUsLinkComponent
10-Benzylamino-1-decanol ≥98.0% (by titrimetric analysis)
Irritant
10-Benzylamino-1-decanol ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2386-5G
Supplier:  TCI
CAS Number:  
Irritant
10-Benzylamino-1-decanol ≥98.0% (by titrimetric analysis)
Catalog # TCIAB2386-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid to slightly cloudy liquid
  • Purity (GC):
    min. 96.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

Specification Test Results

About this item

 :
 :263.43 g/mol
 :GHS07
CAS Number:1274892-05-5
Formula:C₁₇H₂₉NO
MDL Number:MFCD06797073