1,2-Bis(3-indenyl)ethane ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:18657-57-3
:
Formula:C₂₀H₁₈
MDL Number:MFCD00191814
:258.36 g/mol
Specification Test Results
- Appearance:White to Yellow powder to crystal
- Purity (HPLC):min. 98.0 area%
- Melting point:121.0 to 126.0 deg-C