
(R,R)-2,6-Bis(4-isopropyl-2-oxazolin-2-yl)pyridine ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:131864-67-0
:
Formula:C₁₇H₂₃N₃O₂
MDL Number:MFCD01321334
:301.39 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Optical purity (LC):min. 98.0 ee%
- Melting point:152.0 to 156.0 deg-C
Specification Test Results
Frequently Bought Together