Nɑ,N-ε-Bis(tert-butoxycarbonyl)-L-lysine dicyclohexylammonium salt ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15098-69-8
:
Formula:C₂₈H₅₃N₃O₆
MDL Number:MFCD00038892
:527.75 g/mol
EINECS:239-150-9
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:139.0 to 143.0 deg-C
- Specific rotation [a]20/D:-4.0 to -8.0 deg (C=3, AcOH)