4-[(tert-Butoxycarbonylamino)methyl]pyridine-2-carboxylic acid ≥96.0% (by HPLC, titration analysis) reagent for application of the exciton chirality method
Supplier: TCI
Certificates
About this item
CAS Number:260970-57-8
:
Formula:C₁₂H₁₆N₂O₄
MDL Number:MFCD02093452
:252.27 g/mol
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (HPLC):min. 96.0 area%
- Purity (Neutralization titration):min. 96.0 %