
(+)-Bicuculline ≥98.0% (by HPLC, titration analysis)
TCIAB1890-25MG
: TCI
:
- Förp.:25 mg
- Seq:2315
- Appearance:White to Yellow to Orange powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:195.0 to 199.0 deg-C
- Specific rotation [a]20/D:+127.0 to +132.0 deg (C=1, CHCl3)
:
:14,01203
:6.1,III
:367.36 g/mol
:207-619-7
:1544
:GHS06
:485-49-4
:C₂₀H₁₇NO₆
:MFCD00005006