Order Entry
Sweden
ContactUsLinkComponent
N-Benzylcinchoninium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
Irritant
N-Benzylcinchoninium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
Catalog # TCIAB1689-5G
Supplier:  TCI
CAS Number:  
Irritant
N-Benzylcinchoninium chloride ≥98.0% (by titrimetric analysis) chiral phase-transfer catalyst
Catalog # TCIAB1689-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystaline
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    +167 to +174 deg (C=0.4, MeOH:H2O=1:5)

Specification Test Results

About this item

 :
 :420.98 g/mol
EINECS:273-915-8
 :GHS07
CAS Number:69221-14-3
Formula:C₂₆H₂₉ClN₂O
MDL Number:MFCD00082423