Order Entry
Sweden
ContactUsLinkComponent
N-Benzylcinchonidinium chloride ≥98.0% (by HPLC, titration analysis) chiral phase-transfer catalyst
N-Benzylcinchonidinium chloride ≥98.0% (by HPLC, titration analysis) chiral phase-transfer catalyst
Catalog # TCIAB1683-10G
Supplier:  TCI
CAS Number:  
N-Benzylcinchonidinium chloride ≥98.0% (by HPLC, titration analysis) chiral phase-transfer catalyst
Catalog # TCIAB1683-10G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Specific rotation [a]20/D:
    -182.0 to -188.0 deg (C=1, H2O)

Specification Test Results

About this item

 :
 :420.98 g/mol
EINECS:273-938-3
 :GHS07
CAS Number:69257-04-1
Formula:C₂₆H₂₉ClN₂O
MDL Number:MFCD00082422