
Bis(8-quinolinolato)zinc(II) hydrate ≥93.0%
Supplier: TCI
Certificates
About this item
CAS Number:13978-85-3
:
Formula:C₁₈H₁₂N₂O₂Zn
MDL Number:MFCD00065291
:353.69 g/mol
EINECS:237-762-0
:GHS07
Specification Test Results
- Appearance:Yellow to Brown to Dark green powder to crystal
- Purity (Chelometric Titration):min. 93.0 %
Specification Test Results
Frequently Bought Together