Order Entry
Sweden
ContactUsLinkComponent
 
 
Butyl-2-bromopropionate ≥95.0% (by GC)
  TCIAB1031-25ML
 :  TCI
 :  
 
Butyl-2-bromopropionate ≥95.0% (by GC)
  TCIAB1031-25ML
 :  TCI
 :  

 

  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :209.08 g/mol
 :196°C
 :255-241-6
 :GHS07
 :41145-84-0
 :C₇H₁₃BrO₂
 :MFCD00059469