Order Entry
Sweden
ContactUsLinkComponent
N-(4-Butoxybenzylidene)-4-acetylaniline ≥98.0% (by titrimetric analysis)
Irritant
N-(4-Butoxybenzylidene)-4-acetylaniline ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0372-1G
Supplier:  TCI
CAS Number:  
Irritant
N-(4-Butoxybenzylidene)-4-acetylaniline ≥98.0% (by titrimetric analysis)
Catalog # TCIAB0372-1G
Supplier:  TCI
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange powder to crystal
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Methanol:
    almost transparency

Specification Test Results

About this item

 :
 :295.38 g/mol
 :GHS07
CAS Number:17224-18-9
Formula:C₁₉H₂₁NO₂
MDL Number:MFCD00059418